Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Cytokeratin 7 Recombinant Protein Antigen
SDP

Catalog No. NB036501 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036501 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036501 Supplier Novus Biologicals™ Supplier No. NBP288918PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results

Specifications

Gene ID (Entrez) 3855
Protein Family Cytokeratin 7
Purity >80% SDS-PAGE
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Formulation PBS, 1 M Urea, pH 7.4.
Gene Symbol KRT7
Quantity 100 μg
Research Category Cell Biology, Cellular Markers
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-55071. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.