Learn More
Description
Host Species: Mouse
Clone: K8.8 + DC10
Isotype: IgG1/κ + IgG1
Species Reactivity: Human. Others not known.
Immunogen: Semi-purified human cytokeratin preparation
Molecular Weight: 52.5 kDa (Cytokeratin 8) and 45 kDa (Cytokeratin 18)
Positive Control: Skin or Breast CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Cytokeratin 8/18 Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | K8.8 + DC10 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Semi-purified human cytokeratin preparation |
| Quantity | 200 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.