Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 8/18 Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1603R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1603R1 1 mL Prediluted
MS1603P1ABX 100 μL Concentrated
MS1603PABX 200 μL Concentrated
MS1603P1 500 μL Concentrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1603R1 Supplier Epredia Supplier No. MS1603R1

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 8/18 Ab-2, Mouse Monoclonal Antibody.

Keratin 8 belongs to the type B (basic) subfamily of high molecular weight keratins and exists in combination with cytokeratin 18 (type A; acidic subfamily of low molecular weight keratins).

Host Species: Mouse

Clone: K8.8 + DC10

Isotype: IgG1/κ + IgG1

Species Reactivity: Human. Others not known.

Immunogen: Semi-purified human cytokeratin preparation

Molecular Weight: 52.5 kDa (Cytokeratin 8) and 45 kDa (Cytokeratin 18)

Positive Control: Skin or Breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Cytokeratin 8/18 Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone K8.8 + DC10
Concentration Prediluted
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Semi-purified human cytokeratin preparation
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.