Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 8/18 Antibody (B22.1/B23.1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311504IR
Description
Cytokeratin 8/18 Monoclonal antibody specifically detects Cytokeratin 8/18 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Cytokeratin 8/18 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
B22.1/B23.1 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
Cytoskeletal preparation from HeLa epitheliod carcinoma cells (B22.1) and PMC-42 breast carcinoma cells of human origin (B23.1) | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Phospho Specific | |
3856 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction