Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 8/18 Antibody (C-51), Allophycocyanin , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311565APC
Description
Cytokeratin 8/18 Monoclonal antibody specifically detects Cytokeratin 8/18 in Human, Rat, Porcine, Bovine, Ferret (Negative), Monkey, Sheep, Chicken (Negative), Donkey (Negative), Mouse (Negative), Rabbit (Negative), Xenopus (Negative) samples. It is validated for Flow CytometrySpecifications
Cytokeratin 8/18 | |
Monoclonal | |
APC | |
CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
Cytoskeleton preparation from HeLa cells | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Phospho Specific | |
3856 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry | |
C-51 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat, Pig, Bovine, Ferret (Negative), Monkey, Sheep | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction