Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 8/18 Antibody (C-51), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP2449290.02MG
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Cytokeratin 8/18 Monoclonal specifically detects Cytokeratin 8/18 in Human, Rat, Porcine, Bovine, Ferret (Negative), Monkey, Sheep, Chicken (Negative), Donkey (Negative), Mouse (Negative), Rabbit (Negative), Xenopus (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
Cytokeratin 8/18 | |
Monoclonal | |
0.2mg/mL | |
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml | |
CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Cytokeratin 8 (CK8) belongs to the type II (or B or basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 18 (CK18). This MAb recognizes all simple epithelia including glandular epithelium, for example thyroid, female breast, gastrointestinal tract, respiratory tract, and urogenital tract including transitional epithelium. All adenocarcinomas and most squamous carcinomas are positive but keratinizing squamous carcinomas are usually negative. This antibody is useful in demonstrating the presence of Paget cells; there is very little keratin 18 in the normal epidermis so only Paget cells are stained. | |
Store at 4C. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
C-51 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
KRT8 | |
Cytoskeleton preparation from HeLa cells | |
0.02 mg | |
Cancer, Cytoskeleton Markers | |
3856 | |
Human, Rat, Pig, Ferret (Negative), Monkey, Primate, Sheep, Chicken (Negative), Donkey (Negative), Mouse (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction