Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 8/18 Antibody (KRT8.18/1346), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $526.00
Specifications
Antigen | Cytokeratin 8/18 |
---|---|
Clone | KRT8.18/1346 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25333020
![]() |
Novus Biologicals
NBP25333020UG |
20 μg |
Each for $268.00
|
|
|||||
NB253330100
![]() |
Novus Biologicals
NBP253330100UG |
100 μg |
Each for $526.00
|
|
|||||
NB253330200
![]() |
Novus Biologicals
NBP253330200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Cytokeratin 8/18 Monoclonal specifically detects Cytokeratin 8/18 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
Cytokeratin 8/18 | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Cytoskeleton Markers | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
3856 | |
Cytoskeleton preparation from HeLa cells | |
Primary | |
Store at 4C. |
KRT8.18/1346 | |
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
KRT8 | |
IgG1 κ | |
Protein A or G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title