Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 8/18 Antibody (rKRT8.18/1346), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308300AF594
Description
Cytokeratin 8/18 Monoclonal antibody specifically detects Cytokeratin 8/18 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Cytokeratin 8/18 | |
Monoclonal | |
Alexa Fluor 594 | |
CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
Recombinant full-length human KRT8 protein & Recombinant full-length human KRT18 protein | |
100 μg | |
Cancer, Cell Biology, Cellular Markers | |
3856 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
rKRT8.18/1346 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction