Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 8/18 Antibody (SPM141), Alexa Fluor™ 700 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311564AF700
Description
Cytokeratin 8/18 Monoclonal antibody specifically detects Cytokeratin 8/18 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Cytokeratin 8/18 | |
Monoclonal | |
Alexa Fluor 700 | |
CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
Keratin preparation from human breast carcinoma MCF-7 cells | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Phospho Specific | |
3856 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SPM141 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction