Learn More
Invitrogen™ Cytokeratin 8 Recombinant Rabbit Monoclonal Antibody (SU0338)

Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Specifications
Specifications
| Antigen | Cytokeratin 8 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | SU0338 |
| Concentration | 1.065 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 40% Glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4 |
| Gene | KRT8 |
| Gene Accession No. | P05787, P11679 |
| Gene Alias | AA960620; AL022697; AU019895; CARD2; cb186; CK8; CK-8; CYK8; CYKER; cytokeratin 8; cytokeratin 8 (370 AA); cytokeratin endo A; Cytokeratin-8; cytokeratin-A; DreK8; EndoA; etID33550.23; etID43142.23; etID43142.8; K2C8; K8; KERA; keratin 8; keratin 8, type II; keratin complex 2, basic, gene 8; keratin, type 2, gene 8; Keratin, type II cytoskeletal 8; keratin, type II cytoskeletal 8-like protein; keratin-8; KO; Krt-2.8; Krt2-8; krt8; LOW QUALITY PROTEIN: keratin, type II cytoskeletal 8; sb:cb186; type II cytokeratin A (no.8); type-II keratin Kb8; wu:fa20h05; wu:fa95h10; wu:fb96c06; zfk8; zf-K8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.