Learn More
Description
Host Species: Mouse
Clone: TS1
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: aa 343-357
Immunogen: Keratin preparation from a human carcinoma
Molecular Weight: 52.5 kDa
Positive Control: MCF-7 or A431 cells; Skin; Colon, Lung or Breast CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunoprecipitation
Specifications
Specifications
| Antigen | Cytokeratin 8 Ab-4 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | TS1 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Keratin preparation from a human carcinoma |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.