Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 8 Ab-4 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS997P1ABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
200µg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS997P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS997P1 500 μL Unconjugated Concentrated 200µg/mL
MS997P 1 mL Unconjugated Concentrated 200μg/mL
MS997R7 7 mL Unconjugated Prediluted Prediluted
MS997B0 100 μL Biotin Concentrated 200μg/mL
MS997B1 500 μL Biotin Concentrated 200μg/mL
MS997B 1 mL Biotin Concentrated 200μg/mL
MS997P0 100 μL Unconjugated Concentrated 200µg/mL
MS997PABX 200 μL Unconjugated Concentrated 1mg/mL
MS997R1 1 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS997P1ABX Supplier Epredia Supplier No. MS997P1ABX

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Ensure accurate, reproducible results in immunohistochemistry and immunoprecipitation experiments with Epredia™ Cytokeratin 8 Ab-4, Mouse Monoclonal Antibody.

Keratin 8 belongs to the type B (basic) subfamily of high molecular weight keratins and exists in combination with keratin 18. Keratin 8 is primarily found in the non-squamous epithelia and is present in majority of adenocarcinomas and ductal carcinomas. It is absent in squamous cell carcinomas. Hepatocellular carcinomas are defined by the use of antibodies that recognize only cytokeratin polypeptides 8 and 18.

Host Species: Mouse

Clone: TS1

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: aa 343-357

Immunogen: Keratin preparation from a human carcinoma

Molecular Weight: 52.5 kDa

Positive Control: MCF-7 or A431 cells; Skin; Colon, Lung or Breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunoprecipitation

Specifications

Antigen Cytokeratin 8 Ab-4
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone TS1
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Keratin preparation from a human carcinoma
Quantity 100 μL
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.