Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 8 Antibody (B22.1), PerCP , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311566PCP
Description
Cytokeratin 8 Monoclonal antibody specifically detects Cytokeratin 8 in Human, Porcine, Bovine, Rabbit, Sheep, Chicken (Negative), Mouse (Negative), Rat (Negative), Xenopus (Negative) samples. It is validated for Flow CytometrySpecifications
Cytokeratin 8 | |
Monoclonal | |
PerCP | |
CARD2, CK8, CK-8, CYK8, Cytokeratin-8, K2C8, K8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
Cytoskeletal preparation from HeLa cells | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Phospho Specific | |
3856 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry | |
B22.1 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Pig, Bovine, Rabbit, Sheep, Chicken (Negative), Mouse (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction