Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 8 Antibody (SPM192), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234400MFV450
Description
Cytokeratin 8 Monoclonal antibody specifically detects Cytokeratin 8 in Human, Rat (Negative) samples. It is validated for Flow CytometrySpecifications
Cytokeratin 8 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat (Negative) | |
Purified |
Flow Cytometry | |
SPM192 | |
Flow Cytometry | |
CARD2, CK8, CK-8, CYK8, Cytokeratin-8, K2C8, K8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
Keratin preparation from a human carcinoma (Uniprot: P05787) | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Phospho Specific | |
3856 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction