Learn More
Invitrogen™ Cytokeratin 8 Monoclonal Antibody (LP3K), eBioscience™

Description
This LP3K monoclonal antibody reacts with human cytokeratin 8 (K8), a 55-kDa member of the family of intermediate filament proteins. Cytokeratin 8 is a type II (or basic) keratin that is expressed in epithelial and carcinoma cells. Cytokeratins form the intracellular cytoskeletal network that maintains the integrity and stability of cells and tissues. More specifically, studies have demonstrated the involvement of cytokeratin 8 in protection against apoptosis, stress, or injury, as well as regulation of the cell cycle. This keratin is frequently co-expressed with cytokeratin 18, a type I (or acidic) keratin as a heterodimer. Although detected primarily in the cytoplasm of normal healthy cells, cytokeratin 8 has been found to localize to the plasma membrane in some tumor cells. Finally, cytokeratin 8 is phosphorylated on serine 73 in dividing cells. Applications Reported: This LP3K antibody has been reported for use in immunoblotting (WB) and immunocytochemistry. Applications Tested: This LP3K antibody has been tested by western blot analysis of reduced A549 cell lysate, as well as by immunocytochemistry of A549 cells. For western blotting, this antibody can be used at 1-10 μg/mL. For immunocytochemistry, LP3K can be used at 1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtrat...
Specifications
Specifications
| Antigen | Cytokeratin 8 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | LP3K |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | KRT8 |
| Gene Accession No. | P05787 |
| Gene Alias | AA960620; AL022697; AU019895; CARD2; cb186; CK8; CK-8; CYK8; CYKER; cytokeratin 8; cytokeratin 8 (370 AA); cytokeratin endo A; Cytokeratin-8; cytokeratin-A; DreK8; EndoA; etID33550.23; etID43142.23; etID43142.8; K2C8; K8; KERA; keratin 8; keratin 8, type II; keratin complex 2, basic, gene 8; keratin, type 2, gene 8; Keratin, type II cytoskeletal 8; keratin, type II cytoskeletal 8-like protein; keratin-8; KO; Krt-2.8; Krt2-8; krt8; LOW QUALITY PROTEIN: keratin, type II cytoskeletal 8; sb:cb186; type II cytokeratin A (no.8); type-II keratin Kb8; wu:fa20h05; wu:fa95h10; wu:fb96c06; zfk8; zf-K8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.