Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 8 Mouse anti-Human, Unconjugated, Clone: 35betaH11, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | Cytokeratin 8 |
---|---|
Clone | 35betaH11 |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABT15292MI
|
MilliporeSigma
MABT152925UG |
25 μg |
Each for $189.29
|
|
|||||
MABT15291MI
|
MilliporeSigma
MABT1529100UG |
100 μg |
Each for $477.80
|
|
|||||
Description
Cytokeratin 8 Monoclonal antibody specifically detects Cytokeratin 8 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blotting.Specifications
Cytokeratin 8 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Keratin;type II cytoskeletal 8;CK-8;Keratin-8;K8;Type-II keratin Kb8 | |
A cytoskeletal preparation of the human hepatocellular carcinoma cell line Hep3B. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
53.70 kDa calculated. |
35betaH11 | |
Monoclonal | |
Purified | |
Cell Structure | |
NP_002264.1 | |
KRT8, CYK8 | |
IgM κ | |
Protein G Purified | |
Clone 35betaH11 is a mouse monoclonal antibody that specifically detetcts human Cytokeratin 8. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title