Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528985 Supplier Invitrogen™ Supplier No. PA528985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, mouse liver. Predicted reactivity: Mouse (87%), Rat (88%), Dog (81%), Pig (85%), Rhesus Monkey (98%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Cytokeratin 8 is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. Cutokeratin 8 typically dimerizes with keratin 18 to form an intermediate filament in simple single-layered epithelial cells. This protein plays a role in maintaining cellular structural integrity and also functions in signal transduction and cellular differentiation. Mutations in this gene cause cryptogenic cirrhosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 8
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene KRT8
Gene Accession No. P05787, P11679
Gene Alias AA960620; AL022697; AU019895; CARD2; cb186; CK8; CK-8; CYK8; CYKER; cytokeratin 8; cytokeratin 8 (370 AA); cytokeratin endo A; Cytokeratin-8; cytokeratin-A; DreK8; EndoA; etID33550.23; etID43142.23; etID43142.8; K2C8; K8; KERA; keratin 8; keratin 8, type II; keratin complex 2, basic, gene 8; keratin, type 2, gene 8; Keratin, type II cytoskeletal 8; keratin, type II cytoskeletal 8-like protein; keratin-8; KO; Krt-2.8; Krt2-8; krt8; LOW QUALITY PROTEIN: keratin, type II cytoskeletal 8; sb:cb186; type II cytokeratin A (no.8); type-II keratin Kb8; wu:fa20h05; wu:fa95h10; wu:fb96c06; zfk8; zf-K8
Gene Symbols KRT8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Cytokeratin 8. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16691, 3856
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.