Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 8 Polyclonal Antibody, Huabio

Catalog No. 502544724
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4724 Supplier HUABIO RESEARCH INC Supplier No. ER180243
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytokeratin 8 Polyclonal antibody specifically detects Cytokeratin 8 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Cytokeratin 8
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene KRT8
Gene Accession No. P05787, P11679, Q10758
Gene Alias AA960620; AL022697; AU019895; CARD2; cb186; CK8; CK-8; CYK8; CYKER; cytokeratin 8; cytokeratin 8 (370 AA); cytokeratin endo A; Cytokeratin-8; cytokeratin-A; DreK8; EndoA; etID33550.23; etID43142.23; etID43142.8; K2C8; K8; KERA; keratin 8; keratin 8, type II; keratin complex 2, basic, gene 8; keratin, type 2, gene 8; Keratin, type II cytoskeletal 8; keratin, type II cytoskeletal 8-like protein; keratin-8; KO; Krt-2.8; Krt2-8; krt8; LOW QUALITY PROTEIN: keratin, type II cytoskeletal 8; sb:cb186; type II cytokeratin A (no.8); type-II keratin Kb8; wu:fa20h05; wu:fa95h10; wu:fb96c06; zfk8; zf-K8
Host Species Rabbit
Immunogen Synthetic peptide within N-terminal human Cytokeratin 8.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16691, 25626, 3856
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.