Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cytokeratin 81/86 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104293
Description
Antibody detects endogenous levels of total KRT81/86.
The protein encoded by this gene is a member of the keratin gene family. As a type II hair keratin, it is a basic protein which heterodimerizes with type I keratins to form hair and nails. The type II hair keratins are clustered in a region of chromosome 12q13 and are grouped into two distinct subfamilies based on structure similarity. One subfamily, consisting of KRTHB1, KRTHB3, and KRTHB6, is highly related. The other less-related subfamily includes KRTHB2, KRTHB4, and KRTHB5. All hair keratins are expressed in the hair follicle; this hair keratin, as well as KRTHB3 and KRTHB6, is found primarily in the hair cortex. Mutations in this gene and KRTHB6 have been observed in patients with a rare dominant hair disease, monilethrix.
Specifications
Cytokeratin 81/86 | |
Polyclonal | |
Unconjugated | |
KRT86 | |
ghHb1; ghHKb1; hair keratin K2.11; Hair keratin K2.9; hard keratin, type II, 1; hard keratin, type II, 6; Hb1; Hb-1; HB6; hHAKB2-1; K81; K86; keratin 81; keratin 81, type II; keratin 86; keratin 86, type II; keratin protein HB6; keratin, hair, basic, 1; keratin, hair, basic, 6 (monilethrix); keratin, type II cuticular Hb1; Keratin, type II cuticular Hb6; keratin-81; Keratin-86; KRT81; KRT86; KRTHB1; KRTHB6; Metastatic lymph node 137 gene protein; MLN 137; MLN137; MNX; Type II hair keratin Hb1; type II hair keratin Hb6; Type-II keratin Kb21; type-II keratin Kb26 | |
Rabbit | |
Affinity chromatography | |
RUO | |
16679, 3887, 3892, 64818 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O43790, P97861, Q14533, Q9ERE2 | |
KRT81, KRT86 | |
A synthesized peptide derived from human KRT86(Accession O43790), corresponding to amino acid residues V261-H311. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction