Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 85/86 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100851
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100851 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100851 Supplier Invitrogen™ Supplier No. PA5100851
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KRT85/86.

The protein encoded by this gene is a member of the keratin gene family. As a type II hair keratin, it is a basic protein which heterodimerizes with type I keratins to form hair and nails. The type II hair keratins are clustered in a region of chromosome 12q13 and are grouped into two distinct subfamilies based on structure similarity. One subfamily, consisting of KRTHB1, KRTHB3, and KRTHB6, is highly related. The other less-related subfamily includes KRTHB2, KRTHB4, and KRTHB5. All hair keratins are expressed in the hair follicle; this hair keratin, as well as KRTHB1 and KRTHB6, is found primarily in the hair cortex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 85/86
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KRT86
Gene Accession No. O43790, P78386, P97861, Q9Z2T6
Gene Alias 5430421N21Rik; AA589597; AI325212; BB007925; ECTD4; hair keratin K2.10; Hair keratin K2.11; hair keratin K2.12; hard keratin, type II, 3; hard keratin, type II, 5; hard keratin, type II, 6; Hb1; HB3; Hb-3; Hb5; Hb-5; HB6; hHb3; hHb5; K83; K85; K86; keratin 83; keratin 83, type II; keratin 85; keratin 85, type II; keratin 86; keratin 86, type II; keratin complex 2, basic, gene 10; keratin complex 2, basic, gene 18; keratin protein HB6; keratin type II; keratin, hair, basic, 3; keratin, hair, basic, 5; keratin, hair, basic, 6 (monilethrix); keratin, type II cuticular Hb3; Keratin, type II cuticular Hb5; keratin, type II cuticular Hb6; Keratin-83; keratin-85; Keratin-86; Khb4; Krt2-10; Krt2-11; Krt2-18; KRT83; Krt85; KRT86; KRTHB1; KRTHB3; KRTHB5; KRTHB6; MHb4; MNX; RIKEN cDNA 5430421N21 gene; type II hair keratin; type II hair keratin Hb3; Type II hair keratin Hb5; type II hair keratin Hb6; Type-II keratin Kb23; type-II keratin Kb25; Type-II keratin Kb26
Gene Symbols KRT85, KRT86
Host Species Rabbit
Immunogen A synthesized peptide derived from human KRT85(Accession P78386), corresponding to amino acid residues R307-K357.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16679, 3891, 3892, 53622
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.