Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin, Acidic (Type I or LMW) Antibody (AE1), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262221APC
Description
Cytokeratin, Acidic (Type I or LMW) Monoclonal specifically detects Cytokeratin, Acidic (Type I or LMW) in Human, Mouse, Rat, Canine, Chicken, Feline, Primate, Rabbit samples. It is validated for Flow Cytometry.Specifications
Cytokeratin, Acidic (Type I or LMW) | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
Primary | |
Human, Mouse, Rat, Canine, Chicken, Feline, Primate, Rabbit | |
IgG1 |
Flow Cytometry | |
AE1 | |
Flow Cytometry | |
CK10, CK14, CK15, CK16, CK19, cytokeratin 10, cytokeratin 14, cytokeratin 15, cytokeratin 16, cytokeratin 19 | |
Human epidermal keratins | |
0.1 mL | |
The antibody MEM-181 reacts with CD25 (Interleukin-2 receptor alpha chain), a 55 kDa type I transmembrane glycoprotein expressed on activated B and T lymphocytes, activated monocytes/macrophages and on CD4+ T lymphocytes (T regulatory cells); it is lost on resting B and T lymphocytes. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction