Learn More
Epredia™ Lab Vision™ Cytokeratin, High Molecular Weight Ab-3, Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: 34βE12
Isotype: IgG1
Species Reactivity: Human, Monkey, Horse and Rabbit. Others not known.
Epitope: Not determined
Immunogen: Solubilized keratin extracted from human stratum corneum
Molecular Weight: 68 kDa (Cytokeratin 1), 58 kDa (Cytokeratin 5), 56.5 kDa (Cytokeratin 10) and 50 kDa (Cytokeratin 14)
Positive Control: Squamous cell CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
Specifications
| Antigen | Cytokeratin, High Molecular Weight Ab-3 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 34BetaE12 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Solubilized keratin extracted from human stratum corneum |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Research Discipline | Cytoskeletal Protein |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.