Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cytokeratin, High Molecular Weight Ab-3, Mouse Monoclonal Antibody

Catalog No. MS1447S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1447S1 500 μL Concentrated
MS1447S0 100 μL Concentrated
MS1447S 1 mL Concentrated
MS1447R7 7 mL Prediluted
MS1447RQ 12 mL Ready-to-Use
MS1447R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS1447S1 Supplier Epredia™ Supplier No. MS1447S1

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

In normal cells, Ab-3 labels squamous, ductal and other complex epithelia. It reacts with benign small-acinir lesions of the prostate and does not react with hepatocytes, pancreatic acinar cells, proximal renal tubes or endometrial glands. In tumor cells, this antibody is reactive with both squamous and ductal neoplasms and variably with those derived from simple epithelium.

Host Species: Mouse

Clone: 34βE12

Isotype: IgG1

Species Reactivity: Human, Monkey, Horse and Rabbit. Others not known.

Epitope: Not determined

Immunogen: Solubilized keratin extracted from human stratum corneum

Molecular Weight: 68 kDa (Cytokeratin 1), 58 kDa (Cytokeratin 5), 56.5 kDa (Cytokeratin 10) and 50 kDa (Cytokeratin 14)

Positive Control: Squamous cell CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Specifications

Antigen Cytokeratin, High Molecular Weight Ab-3
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 34BetaE12
Conjugate Unconjugated
Host Species Mouse
Immunogen Solubilized keratin extracted from human stratum corneum
Quantity 500 μL
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Equine, Human, Non-human Primate, Rabbit
Form Concentrated
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.