Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin, HMW Antibody (AE-3), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP233170MFV500
Description
Cytokeratin, HMW Monoclonal antibody specifically detects Cytokeratin, HMW in Human, Mouse, Rat, Bovine, Canine, Chicken, Monkey, Rabbit samples. It is validated for Flow CytometrySpecifications
Cytokeratin, HMW | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
51350 | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry | |
AE-3 | |
Flow Cytometry | |
CK-2P, cytokeratin 2, Cytokeratin-2P, HUMCYT2A, K2P, K76, keratin 76, keratin, type II cytoskeletal 2 oral, Keratin-76, KRT2Bcytokeratin-2P, KRT2Pkeratin 2p, KRT76, Type-II keratin Kb9 | |
Human epidermal keratin (Uniprot: Q01546) | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Bovine, Canine, Chicken, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction