Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin, HMW Antibody (rKRTH/6617), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308675R
Description
Cytokeratin, HMW Monoclonal antibody specifically detects Cytokeratin, HMW in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Cytokeratin, HMW | |
Monoclonal | |
DyLight 550 | |
CK-2P, cytokeratin 2, Cytokeratin-2P, HUMCYT2A, K2P, K76, keratin 76, keratin, type II cytoskeletal 2 oral, Keratin-76, KRT2Bcytokeratin-2P, KRT2Pkeratin 2p, KRT76, Type-II keratin Kb9 | |
Synthetic peptide corresponding to Cytokeratin, HMW residues within aa300-400 of Cytokeratin, HMW (Uniprot: Q01546) | |
100 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
rKRTH/6617 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
51350 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction