Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin (HMW) Monoclonal Antibody (34bE12), Abbomax

Catalog No. 502204343
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4343 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4343 Supplier Abbomax Inc Supplier No. 605340
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Skin Cellular location: Cytoplasmic.

Cytokeratin (HMW) Monoclonal antibody specifically detects Cytokeratin (HMW) in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Cytokeratin (HMW)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 34bE12
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene KRT10
Gene Accession No. P02533, P04264, P13645, P13647
Gene Alias 58 kda cytokeratin; 67 kDa cytokeratin; BCIE; BIE; CK1; CK-1; CK10; CK-10; CK14; CK-14; CK-2P; CK5; CK-5; cytokeratin 1; cytokeratin 10; cytokeratin 14; cytokeratin 2; cytokeratin-1; Cytokeratin-10; Cytokeratin-14; cytokeratin-2P; Cytokeratin-5; DDD; DDD1; EBS2; EBS3; EBS4; EHK; EHK1; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; epidermolytic hyperkeratosis 1; EPPK; hair alpha protein; HUMCYT2A; K1; K10; K14; K2P; K5; K76; keratin 1; keratin 1, type II; keratin 10; keratin 10, type I; keratin 14; keratin 14, type I; keratin 2p; keratin 5; keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 76; keratin 76, type II; keratin, type I cytoskeletal 10; Keratin, type I cytoskeletal 14; keratin, type II cytoskeletal 1; Keratin, type II cytoskeletal 2 oral; keratin, type II cytoskeletal 5; Keratin-1; Keratin-10; keratin-14; Keratin-5; Keratin-76; KPP; KRT1; KRT10; KRT14; KRT1A; KRT2B; KRT2P; KRT5; KRT5A; KRT76; KRTA; NEPPK; NFJ; Type-II keratin Kb1; type-II keratin Kb5; Type-II keratin Kb9
Host Species Mouse
Immunogen A synthetic peptide of Cytokeratin (HMW)
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3848, 3852, 3858, 3861
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.