Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin HMW Mouse anti-Human, Unconjugated, Clone: 34betaE12, MilliporeSigma™

Catalog No. MABT15301MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT15301MI 100 μL
MABT15302MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT15301MI Supplier MilliporeSigma Supplier No. MABT1530100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin HMW Monoclonal antibody specifically detects Cytokeratin HMW in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin HMW
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 34betaE12
Conjugate Unconjugated
Gene Accession No. NP_006112.3, NP_000415.2, NP_000412, NP_000517.2
Gene Alias Keratin HMW;High molecular weight keratin
Gene Symbols KRT1, KRT5, KRT10, KRT14
Host Species Mouse
Immunogen Solubilized keratin protein derived from human stratum corneum.
Molecular Weight of Antigen 66.04, 62.38, 58.83, and 51.56 kDa calculated for Cytokeratin 1, 5, 10, and 14, respectively.
Purification Method Protein G Purified
Quantity 100 μL
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone 34betaE12 is a mouse monoclonal antibody that detects human cytokeratin.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.