Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin, Low Molecular Weight, Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS341P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
Prediluted
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS341P1 500 μL Concentrated 200μg/mL
MS341P0 100 μL Concentrated 200μg/mL
MS341PABX 200 μL Concentrated 1mg/mL
MS341P 1 mL Concentrated 200μg/mL
MS341R7 7 mL Prediluted Prediluted
MS341P1ABX 100 μL Concentrated 1mg/mL
MS341R1 1 mL Prediluted Prediluted
MS341RQ 12 mL Ready-to-Use Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS341P1 Supplier Epredia Supplier No. MS341P1
Add to Cart
Add to Cart

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI <5.7) and basic (pI >6.0) subfamilies. The acidic keratins have molecular weights of 56.5, 55, 51, 50, 50', 48, 46, 45, and 40 kDa. The basic keratins have molecular weights of 65-67, 64, 59, 58, 56 and 52 kDa. Members of acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with cell type, differentiation status and environment. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

Host Species: Mouse

Clone: AE1

Isotype: IgG1

Species Reactivity: Human, Monkey, Cow, Rabbit, Mouse, Rat and Chicken. Others not known.

Epitope: Not determined

Immunogen: Human epidermal Cytokeratin

Positive Control: Skin, Squamous cell CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Specifications

Antigen Cytokeratin, Low Molecular Weight Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AE1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human epidermal keratin
Quantity 500 μL
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Bovine, Chicken, Human, Mouse, Non-human Primate, Rabbit, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.