Learn More
Cytokeratin, Low Molecular Weight, Ab-1 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: AE1
Isotype: IgG1
Species Reactivity: Human, Monkey, Cow, Rabbit, Mouse, Rat and Chicken. Others not known.
Epitope: Not determined
Immunogen: Human epidermal Cytokeratin
Positive Control: Skin, Squamous cell CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
Specifications
| Antigen | Cytokeratin, Low Molecular Weight Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | AE1 |
| Concentration | Prediluted |
| Conjugate | Unconjugated |
| Formulation | Ready-to-use for IHC (UVLP) |
| Host Species | Mouse |
| Immunogen | Human epidermal keratin |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.