Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin Mouse anti-Human, Rat, Clone: KRT/457, Abnova™

Catalog No. 89034740 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-740 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-740 Supplier Abnova Corporation Supplier No. MAB14657
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against native human Cytokeratin.

  • High specificity
  • High reactivity

Specifications

Antigen Cytokeratin
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone KRT/457
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native human Cytokeratin.
Dilution Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined
Formulation In PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. P02538; P04259; P13647; P19013; P48668; P05787; P13645; P13646; P05783
Gene Alias CK4/CYK4/FLJ31692/K4
Gene Symbols KRT4
Host Species Mouse
Immunogen Keratin-enriched preparation from cultured human epithelial cells.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3851/3852/3853/3854/286887/3856/3858/3860/3875
Test Specificity This antibody recognizes Type I and Type II cytokeratins, including CK4, CK5, CK6, CK8, CK10, CK13, and CK18.
Target Species Human, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.