Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin, pan Antibody (7H8C4), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP151537
Description
Cytokeratin, pan Monoclonal specifically detects Cytokeratin, pan in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
pan Cytokeratin | |
Monoclonal | |
Unconjugated | |
KRT1 | |
This Cytokeratin, pan Antibody (7H8C4) was developed against a purified recombinant fragment of CK5 expressed in E. Coli. | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
3848 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
7H8C4 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
Mouse | |
Unpurified | |
RUO | |
Primary | |
Human | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction