Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin, pan Antibody (C-11), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP148348MFV500
Description
Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human, Mouse, Guinea Pig, Mammalia samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
Cytokeratin, pan | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Guinea Pig, Mammalia | |
Purified |
Flow Cytometry | |
C-11 | |
Flow Cytometry, Flow (Intracellular) | |
AEI2;CK1;EHK;EHK1;EPPK;K1;Keratin, type II cytoskeletal 1;KRT1A;NEPPK | |
This Cytokeratin, pan Antibody (C-11) was developed against a keratin-enriched preparation from human epidermoid carcinoma cell line A431. | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
3848 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction