Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin, pan Antibody (K20), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250220AF532
Description
Cytokeratin, pan Monoclonal specifically detects Cytokeratin, pan in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cytokeratin, pan | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
Protein A purified | |
Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
3848 | |
Human | |
IgG2a κ |
Western Blot, Immunohistochemistry | |
K20 | |
Western Blot, Immunohistochemistry | |
KRT1 | |
This Cytokeratin, pan Antibody (K20) was developed against hair keratin. | |
0.1 mL | |
Primary | |
This antibody recognizes cytokeratins of 46, 48, 50, 55, 56, 56.5, 59, 61, 62, 64, 65, 66 and 67 kDa. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction