Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin, pan Antibody (KRT/457), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247669MFV450
Description
Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Guinea Pig, Goat, Monkey samples. It is validated for Flow CytometrySpecifications
Cytokeratin, pan | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Amphibian, Bovine, Guinea Pig | |
Purified |
Flow Cytometry | |
KRT/457 | |
Flow Cytometry | |
AEI2;CK1;EHK;EHK1;EPPK;K1;Keratin, type II cytoskeletal 1;KRT1A;NEPPK | |
Keratin-enriched preparation from cultured human epithelial cells | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
3848 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction