Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin, pan Antibody (KRT/457), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2453090.02MG
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Cytokeratin, pan Monoclonal specifically detects Cytokeratin, pan in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Guinea Pig, Goat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
pan Cytokeratin | |
Monoclonal | |
0.2mg/mL | |
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
P02538, P04259, P13647, P19013, P48668 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. This antibody recognizes acidic (Type I or LMW) and basic (Type II or HMW) cytokeratins, including 59kDa (CK4); 58kDa (CK5); 56kDa (CK6); 52kDa (CK8); 56.5kDa (CK10); 53kDa (CK13) and 45kDa (CK18). This is a broad-spectrum antibody, which has been reported to differentiate epithelial tumors from non-epithelial tumors. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis. | |
Store at 4C. | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
KRT/457 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
KRT1 | |
Keratin-enriched preparation from cultured human epithelial cells | |
0.02 mg | |
Cytoskeleton Markers, Protein Kinase, Signal Transduction | |
3848 | |
Human, Mouse, Rat, Pig, Amphibian, Bovine, Guinea Pig, Goat, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction