Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin, pan Antibody (KRTL/1077 + KRTH/1076), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311443F
Description
Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Cytokeratin, pan | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
KRTL/1077 + KRTH/1076 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
AEI2;CK1;EHK;EHK1;EPPK;K1;Keratin, type II cytoskeletal 1;KRT1A;NEPPK | |
Recombinant human KRT77 and KRT76 protein | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
3848 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction