Learn More
StatLab Cytology Double Funnel Kit
SDP
Catalog No. STLM96520FW
Click to view available options
Includes:
Double Funnel w/ Filter Card and Cap
Double Funnel w/ White Filter Card, Cap and Clip
Single Funnel w/ Filter Card and Cap
Single Funnel w/ White Filter Card, Cap and Clip
Disposable funnel offers a lower risk of contamination than reusable funnels.
  • Pre-attached filter card for reliable and consistent results
  • For sample volumes up to 0.5 mL
  • Can be used with all stainless steel slide clips
  • 500 per case
Specifications
Capacity (Metric) 0.5 mL
Material Polypropylene
Color Clear
Includes Double Funnel w/ White Filter Card, Cap and Clip
Product Type Cytology Funnel
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.