Learn More
Cytomegalovirus 65 kDa Late Antigen Monoclonal Antibody (B26G)

Description
Cytomegalovirus 65 kDa Late Antigen Monoclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | Cytomegalovirus 65 kDa Late Antigen |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | B26G |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | CMV |
| Host Species | Mouse |
| Immunogen | CMV 65 kDa matrix protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.