Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytomegalovirus 65 kDa Late Antigen Monoclonal Antibody (B37H)
GREENER_CHOICE

Catalog No. PIMA17294
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17294 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17294 Supplier Invitrogen™ Supplier No. MA17294
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7294 antibody reacts with Cytomegalovirus Late Ag. in viral and infected tissue samples. MA1-7294 has been successfully used in ELISA and Immunofluorescence applications. The MA1-7294 antibody was raised against the CMV Late antigen.

Cytomegalovirus (CMV) is an opportunistic pathogen infecting lung, kidney, gut and other organs in situations where an individual is immunologically immature, such as the foetus and neonate. Infection also occurs in immunosuppressed patients e.g. transplant patients, patients undergoing chemotherapy and those with HIV infection. Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytomegalovirus 65 kDa Late Antigen
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone B37H
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias CMV
Host Species Mouse
Immunogen CMV Late antigen.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.