Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytomegalovirus Antibody (1/42) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | Cytomegalovirus |
|---|---|
| Clone | 15342 |
| Concentration | 1.0 mg/mL |
| Dilution | ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry, Immunocytochemistry, ELISA, Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017535
![]() |
Novus Biologicals
NB100630470.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10063047
![]() |
Novus Biologicals
NB10063047 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Cytomegalovirus Monoclonal specifically detects Cytomegalovirus in Cytomegalovirus, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| Cytomegalovirus | |
| 1.0 mg/mL | |
| Immunohistochemistry (Frozen), Immunohistochemistry, Immunocytochemistry, ELISA, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Virus, Cytomegalovirus (CMV) | |
| Partially purified extracts of strain AD169 infected HEL fibroblasts | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 15342 | |
| ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| CMV; HHV 5; HHV5; Human herpesvirus 5 | |
| IgG1 | |
| Protein A or G purified | |
| NB100-63047 is specific for human Cytomegalovirus and can be used to detect an early antigen 6-12 hours post infection. It does not react with laboratory strains of Adenovirus type 7 or other human Herpes viruses. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title