Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytomegalovirus Antibody - BSA Free, Novus Biologicals™

Goat Polyclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | Cytomegalovirus |
|---|---|
| Concentration | 4.0-5.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry (Frozen), ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017689
![]() |
Novus Biologicals
NB100650550.1ML |
0.1 mL |
Each for $206.50
|
|
|||||
NB10065055
![]() |
Novus Biologicals
NB10065055 |
0.5 mL |
Each for $499.50
|
|
|||||
Description
Cytomegalovirus Polyclonal specifically detects Cytomegalovirus in Cytomegalovirus, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| Cytomegalovirus | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline with 0.1% Sodium Azide | |
| CMV; HHV 5; HHV5; Human herpesvirus 5 | |
| Purified virons of strain AD169. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 4.0-5.0 mg/mL | |
| Western Blot, Immunohistochemistry (Frozen), ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| Unconjugated | |
| Goat | |
| Virus, Cytomegalovirus (CMV) | |
| P06473 | |
| gB | |
| IgG | |
| IgG purified | |
| Predominant reactivity is to glycoprotein B. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title