Learn More
Cytomegalovirus Antibody (HCMV16 (51C1)), Alexa Fluor™ 750, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NB10064398AF750
Catalog No. N1064398A70
Only null left
Add to Cart
Description
Description
Cytomegalovirus Monoclonal specifically detects Cytomegalovirus in Cytomegalovirus, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
Specifications
Cytomegalovirus
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
Mouse
Protein A purified
Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
NB100-64398 is specific for Cytomegalovirus glycoprotein H (gH) late neutralizing antigen and reacts with MRC-5 infected cells. It does not react with HSV, VZV, EBV or uninfected cells.
Store at 4°C in the dark.
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
HCMV16 (51C1)
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
CMV; HHV 5; HHV5; Human herpesvirus 5
Cytomegalovirus extracts
0.1 mL
Primary
Cytomegalovirus (CMV), Virus
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.