Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytomegalovirus gB Antibody (hCMV34) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | Cytomegalovirus gB |
|---|---|
| Clone | hCMV34 |
| Concentration | 1.0 mg/mL |
| Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry, Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017597
![]() |
Novus Biologicals
NB100645320.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10064532
![]() |
Novus Biologicals
NB10064532 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Cytomegalovirus gB Monoclonal specifically detects Cytomegalovirus gB in Cytomegalovirus, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| Cytomegalovirus gB | |
| 1.0 mg/mL | |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Virus, Cytomegalovirus (CMV) | |
| Native | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| hCMV34 | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| CMV, CMV glycoprotein B, CMV glycoprotein GP55, Cytomegalovirus glycoprotein GP55, gB, Glycoprotein B, Glycoprotein GP55, HCMV, HCMV glycoprotein B, HCMV glycoprotein GP55, HHV 5 glycoprotein B, HHV 5 glycoprotein GP55, HHV5 glycoprotein B, HHV5 glycoprotein GP55, Human herpesvirus 5 glycoprotein B, Human herpesvirus 5 glycoprotein GP55, UL55 | |
| IgG2a | |
| Protein G purified | |
| Recognizes cytomegalovirus glycoprotein B, a viral ligand for CD209 which allows capture of viral particles by dendritic cells. This does not react with HSV, VSV or EBV. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title