Learn More
Invitrogen™ Cytomegalovirus gB Late Antigen Monoclonal Antibody (1M12)
Description
MA1-7598 detects HCMV from virus samples. MA1-7598 has been successfully used in ELISA, Western blot, and Immunofluorescent procedures. The MA1-7598 immunogen is HCMV infected cells. Store at 4ºC short term. For long term storage, aliquot and freeze at -80ºC.
Specifications
Specifications
| Antigen | Cytomegalovirus gB Late Antigen |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1M12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.01% sodium azide; pH 7.2 |
| Gene Alias | CMV |
| Host Species | Mouse |
| Immunogen | HCMV infected cells. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.