Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytomegalovirus gB Late Antigen Monoclonal Antibody (I85E)
GREENER_CHOICE

Catalog No. MA17293
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17293 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17293 Supplier Invitrogen™ Supplier No. MA17293
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7293 antibody reacts with Cytomegalovirus gB in viral and infected tissue samples. MA1-7293 has been successfully used in ELISA and immunofluorescence applications. The MA1-7293 antibody was raised against CMV gB antigen. This product is suggested for use in, but has not been tested in the following applications: Western Blot, and Immunohistochemistry.

Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5). Glycoprotein B (gB) is the main antigenic protein within the envelope of HCMV. It is required for HCMV mediated membrane fusion along with the gH/gL complex. The unprocessed precursor of HCMV gB is 906 amino acids in length, with a MW of approximately 100kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytomegalovirus gB Late Antigen
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone I85E
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias CMV
Host Species Mouse
Immunogen CMV gB antigen.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.