Learn More
Invitrogen™ Cytomegalovirus gB Late Antigen Polyclonal Antibody

Description
PA1-7211 detects the cytomegalovirus. PA1-7211 has been successfully used in immunofluorescence procedures. The PA1-7211 immunogen is purified virions of strain AD169.
Specifications
Specifications
| Antigen | Cytomegalovirus gB Late Antigen |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 4-5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | CMV |
| Host Species | Goat |
| Immunogen | Purified virions of strain AD169. |
| Purification Method | IgG Fraction |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.