Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytomegalovirus gB Late Antigen Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581766
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581766 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581766 Supplier Invitrogen™ Supplier No. PA581766
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human cytomegalovirus (HCMV) Glycoprotein B/gB.

Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5). Glycoprotein B (gB) is the main antigenic protein within the envelope of HCMV. It is required for HCMV mediated membrane fusion along with the gH/gL complex. The unprocessed precursor of HCMV gB is 906 amino acids in length, with a MW of approximately 100kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytomegalovirus gB Late Antigen
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias CMV
Host Species Rabbit
Immunogen Recombinant Human cytomegalovirus (HCMV) Glycoprotein B/gB protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.