Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytomegalovirus gB Late Antigen Polyclonal Antibody, HRP
GREENER_CHOICE

Catalog No. PIPA173024
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA173024 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA173024 Supplier Invitrogen™ Supplier No. PA173024
Add to Cart
Add to Cart

Goat Polyclonal Antibody

The PA1-73024 antibody reacts with Cytomegalovirus AD 169. PA1-73024 has been successfully used in ELISA applications. The PA1-73024 antibody was raised against purified virions of strain AD 169 (gB antigen).

Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5). Glycoprotein B (gB) is the main antigenic protein within the envelope of HCMV. It is required for HCMV mediated membrane fusion along with the gH/gL complex. The unprocessed precursor of HCMV gB is 906 amino acids in length, with a MW of approximately 100kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytomegalovirus gB Late Antigen
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 10mg/mL BSA and 0.002% thimerosal
Gene Alias CMV
Host Species Goat
Immunogen Purified virions of strain AD 169 (gB antigen).
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.