Learn More
Invitrogen™ Cytomegalovirus gB Late Antigen Recombinant Rabbit Monoclonal Antibody (38)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA529955
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human CMV Glycoprotein B/gB.
Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5). Glycoprotein B (gB) is the main antigenic protein within the envelope of HCMV. It is required for HCMV mediated membrane fusion along with the gH/gL complex. The unprocessed precursor of HCMV gB is 906 amino acids in length, with a MW of approximately 100kDa.
Specifications
Cytomegalovirus gB Late Antigen | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
ELISA, Western Blot | |
38 | |
Unconjugated | |
CMV | |
Recombinant Human CMV gB/Glycoprotein B protein. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.