Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytomegalovirus gH Antibody (HCMV16), DyLight 550 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320158R
Description
Cytomegalovirus gH Monoclonal antibody specifically detects Cytomegalovirus gH in Cytomegalovirus, Virus samples. It is validated for ELISA, ImmunofluorescenceSpecifications
Cytomegalovirus gH | |
Monoclonal | |
DyLight 550 | |
envelope glycoprotein H, gH, gpUL75, HCMV gH, HHV-5 gH, HHV5 glycoprotein H, Human betaherpesvirus 5 envelope glycoprotein H, Human Cytomegalovirus glycoprotein H, UL75 | |
The original antibody was generated by immunization of mice with purified HCMV strain AD169. | |
0.1 mL | |
Primary | |
Cytomegalovirus (CMV), Virus | |
Purified |
ELISA, Immunofluorescence | |
HCMV16 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
3077450 | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction